The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011531267.2 | XP_011529569.1 | 2 - 3 | 298 | 101 |
GenBank mRNA | BC103893.1 | - | 1 - 2 | 45 | 101 |