Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 38601132-38601387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18A
Entrez Gene ID
253650
Gene Name
ankyrin repeat domain 18A
Gene Aliases
-
UniGene
Hs.644630
Gene Chromosome Location
Chr.9: 38566260-38620587 on Build GRCh38
Transcripts
NM_147195.2 XM_006716749.3 XM_011517836.2 XM_011517837.2 XM_011517839.2 XM_017014576.1 XM_017014577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7272338 c.930G>A p.Q310Q Chr.9: 38601137-38601137 on Build GRCh38
6330041 c.874C>T p.L292L Chr.9: 38601193-38601193 on Build GRCh38
6013482 c.914G>A p.R305K Chr.9: 38601153-38601153 on Build GRCh38
5694908 c.896delA p.K299fs*32 Chr.9: 38601171-38601171 on Build GRCh38
5414543 c.895A>T p.K299* Chr.9: 38601172-38601172 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752096464 Chr.9: 38601199-38601199 on Build GRCh38
rs749770220 Chr.9: 38601186-38601186 on Build GRCh38
rs766629343 Chr.9: 38601198-38601198 on Build GRCh38
rs967865854 Chr.9: 38601273-38601273 on Build GRCh38
rs188531409 Chr.9: 38601259-38601259 on Build GRCh38
rs778641301 Chr.9: 38601231-38601231 on Build GRCh38
rs755589951 Chr.9: 38601205-38601205 on Build GRCh38
rs974627332 Chr.9: 38601356-38601356 on Build GRCh38
rs536774449 Chr.9: 38601238-38601238 on Build GRCh38
rs750424947 Chr.9: 38601181-38601181 on Build GRCh38
rs1000224278 Chr.9: 38601281-38601281 on Build GRCh38
rs758505854 Chr.9: 38601190-38601190 on Build GRCh38
rs756810837 Chr.9: 38601216-38601216 on Build GRCh38
rs957957003 Chr.9: 38601337-38601337 on Build GRCh38
rs146495784 Chr.9: 38601334-38601334 on Build GRCh38
rs895958063 Chr.9: 38601182-38601182 on Build GRCh38
rs748574915 Chr.9: 38601215-38601215 on Build GRCh38
rs781661314 Chr.9: 38601207-38601207 on Build GRCh38

View Full Product Details