Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70292501-70292774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
UniGene
Hs.744989
Gene Chromosome Location
Chr.1: 70259002-70354977 on Build GRCh38
Transcripts
NM_030816.4 XM_005271234.3 XM_005271235.3 XM_006710929.3 XM_017002413.1 XM_017002414.1 XM_017002415.1 XM_017002416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384598 c.1064G>T p.G355V Chr.1: 70292539-70292539 on Build GRCh38
6324925 c.1074G>T p.L358F Chr.1: 70292529-70292529 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371532355 Chr.1: 70292535-70292535 on Build GRCh38
rs970438843 Chr.1: 70292727-70292727 on Build GRCh38
rs547606073 Chr.1: 70292673-70292673 on Build GRCh38
rs755737433 Chr.1: 70292561-70292561 on Build GRCh38
rs116596363 Chr.1: 70292616-70292616 on Build GRCh38
rs779692426 Chr.1: 70292570-70292570 on Build GRCh38
rs980128409 Chr.1: 70292740-70292740 on Build GRCh38
rs890153742 Chr.1: 70292652-70292652 on Build GRCh38
rs747843565 Chr.1: 70292577-70292577 on Build GRCh38
rs756875899 Chr.1: 70292531-70292531 on Build GRCh38
rs781048818 Chr.1: 70292573-70292587 on Build GRCh38
rs1017750588 Chr.1: 70292665-70292665 on Build GRCh38
rs956289005 Chr.1: 70292703-70292703 on Build GRCh38
rs1031663709 Chr.1: 70292585-70292585 on Build GRCh38
rs1051409244 Chr.1: 70292638-70292638 on Build GRCh38
rs868362057 Chr.1: 70292549-70292549 on Build GRCh38
rs565901462 Chr.1: 70292718-70292718 on Build GRCh38
rs977464709 Chr.1: 70292576-70292576 on Build GRCh38
rs909143407 Chr.1: 70292710-70292710 on Build GRCh38
rs190964018 Chr.1: 70292723-70292723 on Build GRCh38
rs769016661 Chr.1: 70292588-70292588 on Build GRCh38
rs563813543 Chr.1: 70292682-70292682 on Build GRCh38
rs138638427 Chr.1: 70292687-70292687 on Build GRCh38
rs748626410 Chr.1: 70292599-70292599 on Build GRCh38
rs745580833 Chr.1: 70292541-70292541 on Build GRCh38
rs1037548100 Chr.1: 70292527-70292527 on Build GRCh38
rs188994141 Chr.1: 70292619-70292619 on Build GRCh38
rs565754071 Chr.1: 70292584-70292584 on Build GRCh38
rs370600142 Chr.1: 70292662-70292662 on Build GRCh38
rs774892536 Chr.1: 70292594-70292594 on Build GRCh38
rs1007241419 Chr.1: 70292660-70292660 on Build GRCh38
rs947913131 Chr.1: 70292655-70292655 on Build GRCh38

View Full Product Details