Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206549962-206550211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM23
Entrez Gene ID
8745
Gene Name
ADAM metallopeptidase domain 23
Gene Aliases
MDC-3, MDC3
UniGene
Hs.370287
Gene Chromosome Location
Chr.2: 206443544-206621130 on Build GRCh38
Transcripts
NM_003812.3 XM_005246932.3 XM_011512086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4915867 c.879T>C p.G293G Chr.2: 206550106-206550106 on Build GRCh38
1404834 c.919A>G p.N307D Chr.2: 206550146-206550146 on Build GRCh38
1015424 c.878G>A p.G293D Chr.2: 206550105-206550105 on Build GRCh38
6450861 c.933G>A p.T311T Chr.2: 206550160-206550160 on Build GRCh38
1691862 c.898T>A p.Y300N Chr.2: 206550125-206550125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915841794 Chr.2: 206550052-206550052 on Build GRCh38
rs372261190 Chr.2: 206550159-206550159 on Build GRCh38
rs977657316 Chr.2: 206549997-206549997 on Build GRCh38
rs751138148 Chr.2: 206550062-206550062 on Build GRCh38
rs777098022 Chr.2: 206550170-206550170 on Build GRCh38
rs544891017 Chr.2: 206550015-206550015 on Build GRCh38
rs1037372200 Chr.2: 206550157-206550157 on Build GRCh38
rs749004991 Chr.2: 206550097-206550097 on Build GRCh38
rs758123170 Chr.2: 206550059-206550059 on Build GRCh38
rs190145861 Chr.2: 206550025-206550025 on Build GRCh38
rs774697917 Chr.2: 206550112-206550112 on Build GRCh38
rs1032641140 Chr.2: 206550026-206550026 on Build GRCh38
rs201146986 Chr.2: 206550102-206550102 on Build GRCh38
rs968300265 Chr.2: 206550022-206550022 on Build GRCh38
rs754105212 Chr.2: 206550055-206550055 on Build GRCh38
rs780624220 Chr.2: 206550074-206550074 on Build GRCh38
rs988514911 Chr.2: 206550073-206550073 on Build GRCh38
rs756746305 Chr.2: 206550065-206550065 on Build GRCh38
rs759816576 Chr.2: 206550174-206550174 on Build GRCh38
rs762067461 Chr.2: 206550125-206550125 on Build GRCh38
rs766777709 Chr.2: 206550168-206550168 on Build GRCh38
rs554601426 Chr.2: 206549992-206549992 on Build GRCh38
rs375863378 Chr.2: 206550091-206550091 on Build GRCh38
rs991429737 Chr.2: 206550038-206550038 on Build GRCh38
rs763795167 Chr.2: 206550176-206550176 on Build GRCh38
rs751228343 Chr.2: 206550178-206550178 on Build GRCh38
rs371228884 Chr.2: 206550090-206550090 on Build GRCh38
rs745823546 Chr.2: 206550084-206550084 on Build GRCh38
rs772280362 Chr.2: 206550133-206550133 on Build GRCh38
rs773389657 Chr.2: 206550138-206550138 on Build GRCh38

View Full Product Details