Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70523433-70523660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM20
Entrez Gene ID
8748
Gene Name
ADAM metallopeptidase domain 20
Gene Aliases
-
UniGene
Hs.177984
Gene Chromosome Location
Chr.14: 70520817-70564742 on Build GRCh38
Transcripts
NM_003814.4 XM_005268151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6076172 c.1405G>T p.E469* Chr.14: 70523503-70523503 on Build GRCh38
3815218 c.1265T>G p.V422G Chr.14: 70523643-70523643 on Build GRCh38
158884 c.1336C>T p.Q446* Chr.14: 70523572-70523572 on Build GRCh38
4052096 c.1451G>A p.C484Y Chr.14: 70523457-70523457 on Build GRCh38
1607907 c.1297C>T p.Q433* Chr.14: 70523611-70523611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143888877 Chr.14: 70523606-70523606 on Build GRCh38
rs769342676 Chr.14: 70523478-70523478 on Build GRCh38
rs766018043 Chr.14: 70523496-70523496 on Build GRCh38
rs113521334 Chr.14: 70523589-70523589 on Build GRCh38
rs530466368 Chr.14: 70523487-70523487 on Build GRCh38
rs939926809 Chr.14: 70523461-70523461 on Build GRCh38
rs45622834 Chr.14: 70523631-70523631 on Build GRCh38
rs752606352 Chr.14: 70523528-70523528 on Build GRCh38
rs527303493 Chr.14: 70523530-70523530 on Build GRCh38
rs770508103 Chr.14: 70523596-70523596 on Build GRCh38
rs750539188 Chr.14: 70523563-70523563 on Build GRCh38
rs145788093 Chr.14: 70523561-70523561 on Build GRCh38
rs754969914 Chr.14: 70523510-70523510 on Build GRCh38
rs547855276 Chr.14: 70523490-70523490 on Build GRCh38
rs368537670 Chr.14: 70523565-70523565 on Build GRCh38
rs917509056 Chr.14: 70523580-70523580 on Build GRCh38
rs141923848 Chr.14: 70523515-70523515 on Build GRCh38
rs201314255 Chr.14: 70523609-70523609 on Build GRCh38
rs759240385 Chr.14: 70523501-70523501 on Build GRCh38
rs139798952 Chr.14: 70523632-70523632 on Build GRCh38
rs969232008 Chr.14: 70523509-70523509 on Build GRCh38
rs769252835 Chr.14: 70523572-70523572 on Build GRCh38
rs779521607 Chr.14: 70523585-70523585 on Build GRCh38
rs138880063 Chr.14: 70523567-70523567 on Build GRCh38
rs751610956 Chr.14: 70523551-70523551 on Build GRCh38
rs749707809 Chr.14: 70523568-70523568 on Build GRCh38
rs185813974 Chr.14: 70523497-70523497 on Build GRCh38
rs759107358 Chr.14: 70523605-70523605 on Build GRCh38
rs748686512 Chr.14: 70523594-70523594 on Build GRCh38
rs775126973 Chr.14: 70523608-70523608 on Build GRCh38
rs745796549 Chr.14: 70523488-70523489 on Build GRCh38
rs758174796 Chr.14: 70523600-70523600 on Build GRCh38
rs764071114 Chr.14: 70523546-70523546 on Build GRCh38
rs113006830 Chr.14: 70523508-70523508 on Build GRCh38
rs113741808 Chr.14: 70523559-70523559 on Build GRCh38
rs76176529 Chr.14: 70523570-70523570 on Build GRCh38
rs147557544 Chr.14: 70523479-70523479 on Build GRCh38

View Full Product Details