The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172250.2 | NP_758454.1 | 6 - 7 | 1051 | 110 |
XM_011531684.2 | XP_011529986.1 | 6 - 7 | 1129 | 110 | |
XM_011531685.2 | XP_011529987.1 | 6 - 7 | 1172 | 110 | |
XM_011531686.2 | XP_011529988.1 | 5 - 6 | 1319 | 110 | |
GenBank mRNA | AK126662.1 | - | 6 - 7 | 1051 | 110 |
BC101178.2 | - | 5 - 6 | 1013 | 110 | |
BC101179.2 | - | 5 - 6 | 1013 | 110 | |
BC101180.2 | - | 4 - 5 | 927 | 110 | |
BC101181.2 | - | 5 - 6 | 1013 | 110 |