The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030961.2 | NP_112223.1 | 1 - 2 | 132 | 155 |
GenBank mRNA | BC005847.2 | - | 1 - 2 | 132 | 155 |
BC011882.2 | - | 1 - 2 | 49 | 155 | |
BC048194.1 | - | 1 - 2 | 66 | 155 |