The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001511.3 | NP_001502.1 | 3 - 4 | 388 | 147 |
NR_046035.1 | 3 - 4 | 411 | 147 | ||
GenBank mRNA | BC011976.1 | - | 3 - 4 | 376 | 147 |
BM554448.1 | - | 3 - 4 | 401 | 147 | |
J03561.1 | - | 3 - 4 | 362 | 147 | |
X12510.1 | - | 3 - 4 | 326 | 147 |