Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21301117-21301360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
UniGene
Hs.126035
Gene Chromosome Location
Chr.14: 21287977-21351316 on Build GRCh38
Transcripts
NM_020366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5656080 c.388T>G p.C130G Chr.14: 21301135-21301135 on Build GRCh38
4571042 c.371T>G p.L124R Chr.14: 21301118-21301118 on Build GRCh38
4049806 c.413G>A p.R138H Chr.14: 21301160-21301160 on Build GRCh38
4751063 c.484A>G p.K162E Chr.14: 21301231-21301231 on Build GRCh38
6353278 c.450C>G p.L150L Chr.14: 21301197-21301197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372471202 Chr.14: 21301201-21301201 on Build GRCh38
rs751684215 Chr.14: 21301179-21301179 on Build GRCh38
rs755600406 Chr.14: 21301184-21301184 on Build GRCh38
rs776488741 Chr.14: 21301148-21301148 on Build GRCh38
rs562265480 Chr.14: 21301170-21301170 on Build GRCh38
rs144585562 Chr.14: 21301197-21301197 on Build GRCh38
rs766604917 Chr.14: 21301174-21301174 on Build GRCh38
rs11628996 Chr.14: 21301190-21301190 on Build GRCh38
rs769524098 Chr.14: 21301280-21301280 on Build GRCh38
rs866277722 Chr.14: 21301214-21301214 on Build GRCh38
rs779601115 Chr.14: 21301246-21301246 on Build GRCh38
rs762804365 Chr.14: 21301137-21301155 on Build GRCh38
rs773026720 Chr.14: 21301286-21301286 on Build GRCh38
rs1016587836 Chr.14: 21301218-21301218 on Build GRCh38
rs750630784 Chr.14: 21301161-21301161 on Build GRCh38
rs566003306 Chr.14: 21301212-21301212 on Build GRCh38
rs761513451 Chr.14: 21301159-21301159 on Build GRCh38
rs200740893 Chr.14: 21301248-21301248 on Build GRCh38
rs368528161 Chr.14: 21301220-21301220 on Build GRCh38
rs972428544 Chr.14: 21301265-21301265 on Build GRCh38
rs761758059 Chr.14: 21301275-21301275 on Build GRCh38
rs778179468 Chr.14: 21301208-21301208 on Build GRCh38
rs971651638 Chr.14: 21301327-21301327 on Build GRCh38
rs776461437 Chr.14: 21301251-21301251 on Build GRCh38
rs756459696 Chr.14: 21301206-21301206 on Build GRCh38
rs1025865898 Chr.14: 21301274-21301274 on Build GRCh38
rs766132168 Chr.14: 21301212-21301213 on Build GRCh38

View Full Product Details