Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7001711-7001936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12
Entrez Gene ID
239
Gene Name
arachidonate 12-lipoxygenase, 12S type
Gene Aliases
12-LOX, 12S-LOX, LOG12
UniGene
Hs.654431
Gene Chromosome Location
Chr.17: 6993791-7010736 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Gene Symbol
ALOX12-AS1
Entrez Gene ID
100506713
Gene Name
ALOX12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.422967
Gene Chromosome Location
Chr.17: 6985123-7012334 on Build GRCh38
Transcripts
NM_000697.2 XM_011523780.2 NR_040089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146519 c.1078C>T p.H360Y Chr.17: 7001728-7001728 on Build GRCh38
6598643 c.1103C>T p.A368V Chr.17: 7001753-7001753 on Build GRCh38
6257479 c.1131G>T p.R377R Chr.17: 7001781-7001781 on Build GRCh38
1679442 c.1115C>A p.A372D Chr.17: 7001765-7001765 on Build GRCh38
1679443 c.1141G>A p.G381R Chr.17: 7001791-7001791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537992981 Chr.17: 7001794-7001794 on Build GRCh38
rs748849528 Chr.17: 7001797-7001797 on Build GRCh38
rs756762345 Chr.17: 7001812-7001812 on Build GRCh38
rs138098217 Chr.17: 7001780-7001780 on Build GRCh38
rs971490697 Chr.17: 7001743-7001743 on Build GRCh38
rs746428889 Chr.17: 7001823-7001823 on Build GRCh38
rs994225660 Chr.17: 7001733-7001733 on Build GRCh38
rs747424629 Chr.17: 7001850-7001850 on Build GRCh38
rs986644357 Chr.17: 7001904-7001904 on Build GRCh38
rs779749856 Chr.17: 7001825-7001826 on Build GRCh38
rs778331954 Chr.17: 7001817-7001817 on Build GRCh38
rs753021260 Chr.17: 7001768-7001768 on Build GRCh38
rs923939578 Chr.17: 7001899-7001899 on Build GRCh38
rs777502068 Chr.17: 7001781-7001781 on Build GRCh38
rs559146443 Chr.17: 7001845-7001845 on Build GRCh38
rs902686258 Chr.17: 7001865-7001865 on Build GRCh38
rs542481780 Chr.17: 7001841-7001841 on Build GRCh38
rs767486999 Chr.17: 7001769-7001769 on Build GRCh38
rs11078660 Chr.17: 7001883-7001883 on Build GRCh38
rs1003761536 Chr.17: 7001763-7001763 on Build GRCh38
rs933987170 Chr.17: 7001902-7001902 on Build GRCh38
rs768865817 Chr.17: 7001852-7001852 on Build GRCh38
rs185957514 Chr.17: 7001853-7001853 on Build GRCh38
rs968186808 Chr.17: 7001873-7001873 on Build GRCh38
rs143493293 Chr.17: 7001741-7001741 on Build GRCh38
rs1042357 Chr.17: 7001742-7001742 on Build GRCh38
rs751558695 Chr.17: 7001760-7001760 on Build GRCh38
rs201220028 Chr.17: 7001770-7001770 on Build GRCh38
rs1022367956 Chr.17: 7001857-7001857 on Build GRCh38
rs528224788 Chr.17: 7001869-7001869 on Build GRCh38

View Full Product Details