Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTC1
Entrez Gene ID
70
Gene Name
actin, alpha, cardiac muscle 1
Gene Aliases
ACTC, ASD5, CMD1R, CMH11, LVNC4
Chromosome Location
Chr.15: 34788096 - 34795726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005159.4 NP_005150.1 5 - 6 1057 116
GenBank mRNA AK056592.1 - 5 - 6 597 116
AK129892.1 - 5 - 6 880 116
AK129902.1 - 5 - 6 880 116
AK290616.1 - 5 - 6 880 116
BC009978.2 - 5 - 6 1019 116
CR541795.1 - 4 - 5 814 116

View Full Product Details