Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95034558-95034823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC4
Entrez Gene ID
10257
Gene Name
ATP binding cassette subfamily C member 4
Gene Aliases
MOAT-B, MOATB, MRP4
UniGene
Hs.508423
Gene Chromosome Location
Chr.13: 95019829-95301550 on Build GRCh38
Transcripts
NM_001301829.1 NM_005845.4 XM_005254025.2 XM_017020319.1 XM_017020321.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6286726 c.3797A>T p.E1266V Chr.13: 95034678-95034678 on Build GRCh38
1629191 c.3867A>G p.K1289K Chr.13: 95034608-95034608 on Build GRCh38
3936382 c.3796G>C p.E1266Q Chr.13: 95034679-95034679 on Build GRCh38
5526055 c.3789A>G p.Q1263Q Chr.13: 95034686-95034686 on Build GRCh38
3732233 c.3844G>A p.A1282T Chr.13: 95034631-95034631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150571031 Chr.13: 95034790-95034791 on Build GRCh38
rs147120244 Chr.13: 95034626-95034626 on Build GRCh38
rs777447817 Chr.13: 95034612-95034612 on Build GRCh38
rs11568704 Chr.13: 95034701-95034701 on Build GRCh38
rs773869967 Chr.13: 95034677-95034677 on Build GRCh38
rs200865691 Chr.13: 95034707-95034707 on Build GRCh38
rs1043750290 Chr.13: 95034754-95034754 on Build GRCh38
rs1134217 Chr.13: 95034661-95034661 on Build GRCh38
rs148827070 Chr.13: 95034699-95034699 on Build GRCh38
rs779070576 Chr.13: 95034600-95034600 on Build GRCh38
rs769378898 Chr.13: 95034637-95034637 on Build GRCh38
rs199715614 Chr.13: 95034632-95034632 on Build GRCh38
rs765385261 Chr.13: 95034778-95034778 on Build GRCh38
rs754094423 Chr.13: 95034598-95034598 on Build GRCh38
rs770411207 Chr.13: 95034684-95034684 on Build GRCh38
rs746655525 Chr.13: 95034681-95034681 on Build GRCh38
rs748268663 Chr.13: 95034606-95034606 on Build GRCh38
rs777625353 Chr.13: 95034702-95034702 on Build GRCh38
rs759876521 Chr.13: 95034776-95034776 on Build GRCh38
rs575081939 Chr.13: 95034700-95034700 on Build GRCh38
rs775336450 Chr.13: 95034742-95034742 on Build GRCh38
rs981112090 Chr.13: 95034617-95034617 on Build GRCh38
rs755198749 Chr.13: 95034695-95034695 on Build GRCh38
rs777051259 Chr.13: 95034770-95034770 on Build GRCh38
rs746753787 Chr.13: 95034623-95034623 on Build GRCh38
rs777781664 Chr.13: 95034748-95034750 on Build GRCh38
rs762615043 Chr.13: 95034657-95034657 on Build GRCh38
rs781020367 Chr.13: 95034718-95034718 on Build GRCh38
rs560796150 Chr.13: 95034750-95034750 on Build GRCh38
rs775106710 Chr.13: 95034650-95034650 on Build GRCh38
rs756956177 Chr.13: 95034708-95034708 on Build GRCh38
rs370348158 Chr.13: 95034631-95034631 on Build GRCh38
rs141716134 Chr.13: 95034613-95034613 on Build GRCh38
rs375925660 Chr.13: 95034599-95034599 on Build GRCh38
rs768286310 Chr.13: 95034749-95034749 on Build GRCh38
rs769480579 Chr.13: 95034729-95034729 on Build GRCh38
rs748745765 Chr.13: 95034744-95034744 on Build GRCh38
rs761265453 Chr.13: 95034757-95034757 on Build GRCh38
rs753955694 Chr.13: 95034682-95034682 on Build GRCh38
rs185212700 Chr.13: 95034680-95034680 on Build GRCh38
rs374563885 Chr.13: 95034721-95034721 on Build GRCh38
rs138512563 Chr.13: 95034660-95034660 on Build GRCh38
rs188936746 Chr.13: 95034691-95034691 on Build GRCh38
rs771388445 Chr.13: 95034767-95034767 on Build GRCh38
rs758328249 Chr.13: 95034610-95034610 on Build GRCh38

View Full Product Details