Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138113569-138113842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1D1
Entrez Gene ID
6718
Gene Name
aldo-keto reductase family 1, member D1
Gene Aliases
3o5bred, CBAS2, SRD5B1
UniGene
Hs.201667
Gene Chromosome Location
Chr.7: 138076432-138118304 on Build GRCh38
Transcripts
NM_001190906.1 NM_001190907.1 NM_005989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5515224 c.858C>A p.I286I Chr.7: 138113692-138113692 on Build GRCh38
3778143 c.893A>G p.D298G Chr.7: 138113727-138113727 on Build GRCh38
7318822 c.884A>T p.E295V Chr.7: 138113718-138113718 on Build GRCh38
6303269 c.877G>T p.E293* Chr.7: 138113711-138113711 on Build GRCh38
3633842 c.930A>T p.E310D Chr.7: 138113764-138113764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199670016 Chr.7: 138113781-138113781 on Build GRCh38
rs762812138 Chr.7: 138113816-138113816 on Build GRCh38
rs17169518 Chr.7: 138113680-138113680 on Build GRCh38
rs569520757 Chr.7: 138113660-138113660 on Build GRCh38
rs759271382 Chr.7: 138113670-138113670 on Build GRCh38
rs1014190147 Chr.7: 138113794-138113794 on Build GRCh38
rs768796075 Chr.7: 138113782-138113782 on Build GRCh38
rs775495443 Chr.7: 138113815-138113815 on Build GRCh38
rs775637709 Chr.7: 138113670-138113671 on Build GRCh38
rs761365306 Chr.7: 138113735-138113735 on Build GRCh38
rs777887391 Chr.7: 138113805-138113805 on Build GRCh38
rs116226492 Chr.7: 138113754-138113754 on Build GRCh38
rs759908186 Chr.7: 138113698-138113698 on Build GRCh38
rs772220040 Chr.7: 138113655-138113655 on Build GRCh38
rs563867200 Chr.7: 138113612-138113612 on Build GRCh38
rs754185940 Chr.7: 138113768-138113768 on Build GRCh38
rs1001190278 Chr.7: 138113731-138113731 on Build GRCh38
rs868842878 Chr.7: 138113722-138113722 on Build GRCh38
rs369902735 Chr.7: 138113763-138113763 on Build GRCh38
rs781077807 Chr.7: 138113609-138113609 on Build GRCh38
rs115087848 Chr.7: 138113770-138113770 on Build GRCh38
rs992217685 Chr.7: 138113625-138113625 on Build GRCh38
rs764125206 Chr.7: 138113706-138113706 on Build GRCh38
rs372957383 Chr.7: 138113788-138113788 on Build GRCh38
rs529014981 Chr.7: 138113672-138113672 on Build GRCh38
rs373252045 Chr.7: 138113679-138113679 on Build GRCh38
rs926599247 Chr.7: 138113617-138113617 on Build GRCh38
rs1051566404 Chr.7: 138113595-138113595 on Build GRCh38
rs769563205 Chr.7: 138113782-138113782 on Build GRCh38
rs532824944 Chr.7: 138113618-138113618 on Build GRCh38
rs959475043 Chr.7: 138113619-138113619 on Build GRCh38
rs1034543165 Chr.7: 138113740-138113740 on Build GRCh38
rs748221887 Chr.7: 138113785-138113785 on Build GRCh38
rs756478701 Chr.7: 138113756-138113756 on Build GRCh38
rs773462187 Chr.7: 138113666-138113666 on Build GRCh38
rs1037616786 Chr.7: 138113623-138113623 on Build GRCh38
rs747159861 Chr.7: 138113807-138113807 on Build GRCh38
rs115194708 Chr.7: 138113752-138113752 on Build GRCh38
rs867431553 Chr.7: 138113639-138113639 on Build GRCh38
rs763044178 Chr.7: 138113696-138113696 on Build GRCh38
rs866247461 Chr.7: 138113717-138113717 on Build GRCh38
rs751071947 Chr.7: 138113732-138113732 on Build GRCh38
rs769722522 Chr.7: 138113809-138113809 on Build GRCh38

View Full Product Details