Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51046918-51047155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
6191623 c.97G>A p.E33K Chr.18: 51047143-51047143 on Build GRCh38
5515671 c.91G>A p.E31K Chr.18: 51047137-51047137 on Build GRCh38
189728 c.97G>T p.E33* Chr.18: 51047143-51047143 on Build GRCh38
30786 c.74G>A p.C25Y Chr.18: 51047120-51047120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029971658 Chr.18: 51046963-51046963 on Build GRCh38
rs372316981 Chr.18: 51047066-51047066 on Build GRCh38
rs955324690 Chr.18: 51046977-51046977 on Build GRCh38
rs200236229 Chr.18: 51047016-51047016 on Build GRCh38
rs142292491 Chr.18: 51047067-51047067 on Build GRCh38
rs762273127 Chr.18: 51047055-51047055 on Build GRCh38
rs772186355 Chr.18: 51046978-51046978 on Build GRCh38
rs774342820 Chr.18: 51047053-51047053 on Build GRCh38
rs963833176 Chr.18: 51046956-51046956 on Build GRCh38
rs907866808 Chr.18: 51047000-51047000 on Build GRCh38
rs768803926 Chr.18: 51047037-51047037 on Build GRCh38
rs876658568 Chr.18: 51047069-51047069 on Build GRCh38
rs754523299 Chr.18: 51046965-51046965 on Build GRCh38
rs587780791 Chr.18: 51047077-51047077 on Build GRCh38
rs973470216 Chr.18: 51047049-51047049 on Build GRCh38
rs770128265 Chr.18: 51047012-51047012 on Build GRCh38
rs864622289 Chr.18: 51047038-51047038 on Build GRCh38
rs950414097 Chr.18: 51047097-51047097 on Build GRCh38
rs369978812 Chr.18: 51046996-51046996 on Build GRCh38
rs886053892 Chr.18: 51047044-51047044 on Build GRCh38
rs748143311 Chr.18: 51046975-51046975 on Build GRCh38
rs773111265 Chr.18: 51046991-51046991 on Build GRCh38
rs779811141 Chr.18: 51046950-51046950 on Build GRCh38
rs754401427 Chr.18: 51047103-51047103 on Build GRCh38
rs778465458 Chr.18: 51047130-51047130 on Build GRCh38
rs376371717 Chr.18: 51047085-51047085 on Build GRCh38
rs762899211 Chr.18: 51047019-51047019 on Build GRCh38
rs281875323 Chr.18: 51047084-51047084 on Build GRCh38
rs749026885 Chr.18: 51047016-51047017 on Build GRCh38
rs761044209 Chr.18: 51047076-51047076 on Build GRCh38
rs757702252 Chr.18: 51047054-51047054 on Build GRCh38
rs778679766 Chr.18: 51046967-51046967 on Build GRCh38
rs876659391 Chr.18: 51047116-51047116 on Build GRCh38
rs748692094 Chr.18: 51046958-51046958 on Build GRCh38

View Full Product Details