Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21564176-21564431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPL
Entrez Gene ID
249
Gene Name
alkaline phosphatase, liver/bone/kidney
Gene Aliases
AP-TNAP, APTNAP, HOPS, TNAP, TNSALP
UniGene
Hs.75431
Gene Chromosome Location
Chr.1: 21508982-21578412 on Build GRCh38
Transcripts
NM_000478.5 NM_001127501.3 NM_001177520.2 XM_005245818.1 XM_006710546.2 XM_017000903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400119 c.636C>G p.I212M Chr.1: 21564204-21564204 on Build GRCh38
4661280 c.612C>T p.I204I Chr.1: 21564180-21564180 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs115328321 Chr.1: 21564363-21564363 on Build GRCh38
rs1021979360 Chr.1: 21564382-21564382 on Build GRCh38
rs764771727 Chr.1: 21564249-21564249 on Build GRCh38
rs575597860 Chr.1: 21564246-21564246 on Build GRCh38
rs751081295 Chr.1: 21564267-21564267 on Build GRCh38
rs746899450 Chr.1: 21564256-21564257 on Build GRCh38
rs1056526225 Chr.1: 21564370-21564370 on Build GRCh38
rs967860138 Chr.1: 21564397-21564397 on Build GRCh38
rs926580098 Chr.1: 21564408-21564408 on Build GRCh38
rs543689443 Chr.1: 21564398-21564398 on Build GRCh38
rs185891899 Chr.1: 21564357-21564357 on Build GRCh38
rs956691878 Chr.1: 21564364-21564364 on Build GRCh38
rs774579331 Chr.1: 21564226-21564226 on Build GRCh38
rs914456845 Chr.1: 21564299-21564299 on Build GRCh38
rs1007692398 Chr.1: 21564339-21564339 on Build GRCh38
rs576289457 Chr.1: 21564392-21564392 on Build GRCh38
rs779958465 Chr.1: 21564247-21564247 on Build GRCh38
rs755479390 Chr.1: 21564329-21564329 on Build GRCh38
rs769903299 Chr.1: 21564211-21564211 on Build GRCh38
rs923990321 Chr.1: 21564361-21564361 on Build GRCh38
rs201739155 Chr.1: 21564216-21564216 on Build GRCh38
rs1014629656 Chr.1: 21564230-21564230 on Build GRCh38
rs748031071 Chr.1: 21564197-21564197 on Build GRCh38
rs1006495850 Chr.1: 21564407-21564407 on Build GRCh38
rs1039311319 Chr.1: 21564348-21564348 on Build GRCh38
rs368955682 Chr.1: 21564236-21564236 on Build GRCh38
rs980151458 Chr.1: 21564409-21564409 on Build GRCh38
rs557323064 Chr.1: 21564386-21564386 on Build GRCh38
rs953191402 Chr.1: 21564289-21564289 on Build GRCh38
rs760066009 Chr.1: 21564227-21564227 on Build GRCh38
rs761144902 Chr.1: 21564247-21564247 on Build GRCh38
rs952278980 Chr.1: 21564208-21564208 on Build GRCh38
rs372656246 Chr.1: 21564256-21564256 on Build GRCh38
rs771097001 Chr.1: 21564221-21564221 on Build GRCh38
rs749544042 Chr.1: 21564217-21564217 on Build GRCh38
rs751394580 Chr.1: 21564237-21564237 on Build GRCh38
rs888025311 Chr.1: 21564404-21564404 on Build GRCh38
rs202047015 Chr.1: 21564224-21564224 on Build GRCh38
rs553051630 Chr.1: 21564298-21564298 on Build GRCh38
rs992595722 Chr.1: 21564411-21564411 on Build GRCh38
rs1045289922 Chr.1: 21564402-21564402 on Build GRCh38
rs573908872 Chr.1: 21564254-21564254 on Build GRCh38
rs947341054 Chr.1: 21564313-21564313 on Build GRCh38

View Full Product Details