Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51991796-51992069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
UniGene
Hs.438918
Gene Chromosome Location
Chr.12: 51951667-51997079 on Build GRCh38
Transcripts
NM_004302.4 NM_020327.3 NM_020328.3 XM_011538966.2 XM_017020199.1 XM_017020200.1 XM_017020201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263065 c.1330C>T p.R444* Chr.12: 51991931-51991931 on Build GRCh38
5664872 c.1407G>T p.L469L Chr.12: 51991885-51991885 on Build GRCh38
1676970 c.1460T>A p.V487D Chr.12: 51991938-51991938 on Build GRCh38
6593219 c.1503G>A p.W501* Chr.12: 51991981-51991981 on Build GRCh38
5916147 c.1484C>T p.P495L Chr.12: 51991962-51991962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756507261 Chr.12: 51991983-51991983 on Build GRCh38
rs750811429 Chr.12: 51991848-51991848 on Build GRCh38
rs777137436 Chr.12: 51991990-51991990 on Build GRCh38
rs185200314 Chr.12: 51991838-51991838 on Build GRCh38
rs750709751 Chr.12: 51991971-51991971 on Build GRCh38
rs370721275 Chr.12: 51991945-51991945 on Build GRCh38
rs778353328 Chr.12: 51991845-51991846 on Build GRCh38
rs534751585 Chr.12: 51991850-51991850 on Build GRCh38
rs992309482 Chr.12: 51991858-51991858 on Build GRCh38
rs777798210 Chr.12: 51992020-51992020 on Build GRCh38
rs368988921 Chr.12: 51991965-51991965 on Build GRCh38
rs373149746 Chr.12: 51991894-51991894 on Build GRCh38
rs879457328 Chr.12: 51992028-51992028 on Build GRCh38
rs577618988 Chr.12: 51991942-51991942 on Build GRCh38
rs374460422 Chr.12: 51992023-51992023 on Build GRCh38
rs778812205 Chr.12: 51991882-51991882 on Build GRCh38
rs565517674 Chr.12: 51991847-51991847 on Build GRCh38
rs748761194 Chr.12: 51991867-51991867 on Build GRCh38
rs756525389 Chr.12: 51991829-51991831 on Build GRCh38
rs752777127 Chr.12: 51991822-51991822 on Build GRCh38
rs551127449 Chr.12: 51992010-51992010 on Build GRCh38
rs143166498 Chr.12: 51991912-51991912 on Build GRCh38
rs12228540 Chr.12: 51991839-51991839 on Build GRCh38
rs929924598 Chr.12: 51992022-51992022 on Build GRCh38
rs768090769 Chr.12: 51991970-51991970 on Build GRCh38
rs148254270 Chr.12: 51991967-51991967 on Build GRCh38
rs752062448 Chr.12: 51991831-51991831 on Build GRCh38
rs532947331 Chr.12: 51991924-51991924 on Build GRCh38
rs201642603 Chr.12: 51991821-51991821 on Build GRCh38
rs922311092 Chr.12: 51991864-51991864 on Build GRCh38
rs762169175 Chr.12: 51991969-51991969 on Build GRCh38
rs368700328 Chr.12: 51992034-51992034 on Build GRCh38
rs753543689 Chr.12: 51992005-51992005 on Build GRCh38
rs761136740 Chr.12: 51991852-51991852 on Build GRCh38
rs749452286 Chr.12: 51992031-51992031 on Build GRCh38
rs754482546 Chr.12: 51992007-51992007 on Build GRCh38
rs368192679 Chr.12: 51992009-51992009 on Build GRCh38
rs754152317 Chr.12: 51991847-51991850 on Build GRCh38
rs760541978 Chr.12: 51991932-51991932 on Build GRCh38
rs770900739 Chr.12: 51992025-51992025 on Build GRCh38
rs867736080 Chr.12: 51991872-51991872 on Build GRCh38
rs768200138 Chr.12: 51991873-51991873 on Build GRCh38
rs370784076 Chr.12: 51992013-51992013 on Build GRCh38
rs757702289 Chr.12: 51992045-51992046 on Build GRCh38
rs115149888 Chr.12: 51991825-51991825 on Build GRCh38
rs372305411 Chr.12: 51992044-51992044 on Build GRCh38
rs772828202 Chr.12: 51991915-51991915 on Build GRCh38
rs758222949 Chr.12: 51992014-51992014 on Build GRCh38
rs763103664 Chr.12: 51991964-51991964 on Build GRCh38
rs955677676 Chr.12: 51991827-51991827 on Build GRCh38

View Full Product Details