Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 125316230-125316503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT4
Entrez Gene ID
79633
Gene Name
FAT atypical cadherin 4
Gene Aliases
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2
UniGene
Hs.563205
Gene Chromosome Location
Chr.4: 125314913-125492932 on Build GRCh38
Transcripts
NM_001291285.1 NM_001291303.1 NM_024582.4 XM_011532236.2 XM_011532237.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671272 c.77G>A p.R26Q Chr.4: 125316488-125316488 on Build GRCh38
4683639 c.27T>A p.T9T Chr.4: 125316438-125316438 on Build GRCh38
3825131 c.76C>T p.R26* Chr.4: 125316487-125316487 on Build GRCh38
203298 c.7T>G p.L3V Chr.4: 125316418-125316418 on Build GRCh38
1671274 c.87G>A p.W29* Chr.4: 125316498-125316498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905269379 Chr.4: 125316343-125316343 on Build GRCh38
rs775725065 Chr.4: 125316425-125316425 on Build GRCh38
rs923518964 Chr.4: 125316344-125316344 on Build GRCh38
rs940850181 Chr.4: 125316299-125316299 on Build GRCh38
rs768846719 Chr.4: 125316442-125316442 on Build GRCh38
rs1051236279 Chr.4: 125316353-125316353 on Build GRCh38
rs188629298 Chr.4: 125316390-125316390 on Build GRCh38
rs760841511 Chr.4: 125316439-125316439 on Build GRCh38
rs780994798 Chr.4: 125316368-125316368 on Build GRCh38
rs909355112 Chr.4: 125316267-125316267 on Build GRCh38
rs778698184 Chr.4: 125316407-125316407 on Build GRCh38
rs776935959 Chr.4: 125316379-125316379 on Build GRCh38
rs752357918 Chr.4: 125316373-125316373 on Build GRCh38
rs372684455 Chr.4: 125316415-125316415 on Build GRCh38
rs182890588 Chr.4: 125316300-125316300 on Build GRCh38
rs562838061 Chr.4: 125316325-125316325 on Build GRCh38
rs1045286711 Chr.4: 125316315-125316315 on Build GRCh38
rs942468629 Chr.4: 125316421-125316421 on Build GRCh38
rs771790673 Chr.4: 125316422-125316422 on Build GRCh38
rs369473520 Chr.4: 125316392-125316392 on Build GRCh38
rs763406390 Chr.4: 125316461-125316461 on Build GRCh38
rs375956663 Chr.4: 125316455-125316455 on Build GRCh38
rs940917679 Chr.4: 125316294-125316294 on Build GRCh38
rs559474596 Chr.4: 125316328-125316328 on Build GRCh38
rs755705320 Chr.4: 125316375-125316375 on Build GRCh38
rs939471227 Chr.4: 125316376-125316376 on Build GRCh38
rs753957854 Chr.4: 125316403-125316403 on Build GRCh38
rs369059252 Chr.4: 125316471-125316471 on Build GRCh38
rs776734172 Chr.4: 125316453-125316453 on Build GRCh38
rs1009458431 Chr.4: 125316456-125316456 on Build GRCh38
rs933668902 Chr.4: 125316352-125316352 on Build GRCh38
rs544718440 Chr.4: 125316311-125316311 on Build GRCh38
rs986477734 Chr.4: 125316265-125316269 on Build GRCh38
rs750838482 Chr.4: 125316460-125316460 on Build GRCh38
rs766800954 Chr.4: 125316468-125316468 on Build GRCh38
rs758678446 Chr.4: 125316365-125316365 on Build GRCh38

View Full Product Details