Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP2B4
Entrez Gene ID
493
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Gene Aliases
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
Chromosome Location
Chr.1: 203626787 - 203744081 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001396.2 NP_001001396.1 13 - 14 3103 71
NM_001684.4 NP_001675.3 13 - 14 3103 71
GenBank mRNA AK026443.1 - 1 - 2 129 71
AK292458.1 - 12 - 13 2442 71
BC140774.1 - 13 - 14 2775 71
BC150293.1 - 12 - 13 2460 71
BX537444.1 - 13 - 14 3330 71
CR749393.1 - 13 - 14 3033 71
CR936708.1 - 13 - 14 3058 71
M25874.1 - 12 - 13 2603 71
M83363.1 - 12 - 13 2603 71

View Full Product Details