The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001396.2 | NP_001001396.1 | 3 - 4 | 1289 | 80 |
NM_001684.4 | NP_001675.3 | 3 - 4 | 1289 | 80 | |
GenBank mRNA | AK292458.1 | - | 2 - 3 | 628 | 80 |
BC140774.1 | - | 3 - 4 | 961 | 80 | |
BC150293.1 | - | 2 - 3 | 646 | 80 | |
BX537444.1 | - | 3 - 4 | 1516 | 80 | |
CR749393.1 | - | 3 - 4 | 1219 | 80 | |
CR936708.1 | - | 3 - 4 | 1243 | 80 | |
M25874.1 | - | 2 - 3 | 789 | 80 | |
M83363.1 | - | 2 - 3 | 789 | 80 |