Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALK
Entrez Gene ID
238
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Gene Aliases
CD246, NBLST3
Chromosome Location
Chr.2: 29192774 - 29921611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004304.4 NP_004295.2 7 - 8 2502 91
GenBank mRNA AB209477.4 - 7 - 8 2039 91
EU788003.1 - 7 - 8 2457 91
EU788004.1 - 7 - 8 2457 91
EU788005.1 - 7 - 8 2457 91
EU788006.1 - 7 - 8 2457 91
EU788007.1 - 7 - 8 2457 91
EU788008.1 - 7 - 8 2457 91
EU788009.1 - 7 - 8 2457 91
EU788010.1 - 7 - 8 2457 91
FJ838786.1 - 7 - 8 2457 91
FJ838787.1 - 7 - 8 2457 91
GU128152.1 - 7 - 8 1550 91
GU128153.1 - 7 - 8 1550 91
GU128154.1 - 7 - 8 1550 91
GU128155.1 - 7 - 8 1550 91
GU128156.1 - 7 - 8 1550 91
GU128157.1 - 7 - 8 1550 91
U62540.1 - 7 - 8 2461 91
U66559.1 - 7 - 8 1698 91

View Full Product Details