Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45406991-45407252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3EAP
Entrez Gene ID
10849
Gene Name
CD3e molecule associated protein
Gene Aliases
ASE-1, ASE1, CAST, PAF49
UniGene
Hs.710495
Gene Chromosome Location
Chr.19: 45406129-45410766 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4510202 c.82C>T p.P28S Chr.19: 45407153-45407153 on Build GRCh38
5550045 c.146C>T p.A49V Chr.19: 45407217-45407217 on Build GRCh38
1304775 c.38C>T p.S13F Chr.19: 45407109-45407109 on Build GRCh38
4494807 c.43C>T p.P15S Chr.19: 45407114-45407114 on Build GRCh38
3104130 c.43delC p.N17fs*19 Chr.19: 45407114-45407114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778576229 Chr.19: 45407128-45407128 on Build GRCh38
rs199924316 Chr.19: 45407095-45407095 on Build GRCh38
rs999334010 Chr.19: 45407121-45407121 on Build GRCh38
rs771308001 Chr.19: 45407145-45407146 on Build GRCh38
rs764067005 Chr.19: 45407210-45407210 on Build GRCh38
rs189677989 Chr.19: 45407050-45407050 on Build GRCh38
rs778183041 Chr.19: 45407107-45407109 on Build GRCh38
rs756102625 Chr.19: 45407109-45407109 on Build GRCh38
rs747492022 Chr.19: 45407118-45407119 on Build GRCh38
rs944394165 Chr.19: 45407012-45407012 on Build GRCh38
rs148542036 Chr.19: 45407182-45407182 on Build GRCh38
rs202143184 Chr.19: 45407096-45407096 on Build GRCh38
rs778804838 Chr.19: 45407147-45407147 on Build GRCh38
rs763874860 Chr.19: 45407112-45407112 on Build GRCh38
rs34721735 Chr.19: 45407192-45407193 on Build GRCh38
rs774483707 Chr.19: 45407186-45407186 on Build GRCh38
rs201181795 Chr.19: 45407114-45407114 on Build GRCh38
rs777101778 Chr.19: 45407158-45407158 on Build GRCh38
rs781498182 Chr.19: 45407218-45407219 on Build GRCh38
rs113842913 Chr.19: 45407021-45407021 on Build GRCh38
rs779440899 Chr.19: 45407137-45407137 on Build GRCh38
rs753794315 Chr.19: 45407228-45407228 on Build GRCh38
rs770088433 Chr.19: 45407172-45407172 on Build GRCh38
rs745536961 Chr.19: 45407133-45407133 on Build GRCh38
rs765547678 Chr.19: 45407102-45407102 on Build GRCh38
rs776024192 Chr.19: 45407062-45407062 on Build GRCh38
rs930564230 Chr.19: 45407023-45407023 on Build GRCh38
rs760576094 Chr.19: 45407208-45407208 on Build GRCh38
rs1033040039 Chr.19: 45407125-45407125 on Build GRCh38
rs144685828 Chr.19: 45407103-45407103 on Build GRCh38
rs769080868 Chr.19: 45407074-45407074 on Build GRCh38
rs747519404 Chr.19: 45407068-45407068 on Build GRCh38
rs748478038 Chr.19: 45407163-45407163 on Build GRCh38
rs769350983 Chr.19: 45407152-45407152 on Build GRCh38
rs117289933 Chr.19: 45407107-45407107 on Build GRCh38
rs757857509 Chr.19: 45407136-45407136 on Build GRCh38
rs1023139849 Chr.19: 45407156-45407156 on Build GRCh38
rs745431319 Chr.19: 45407044-45407044 on Build GRCh38
rs777191558 Chr.19: 45407087-45407087 on Build GRCh38
rs908696323 Chr.19: 45407217-45407217 on Build GRCh38
rs367657846 Chr.19: 45407118-45407118 on Build GRCh38
rs372180318 Chr.19: 45407180-45407180 on Build GRCh38
rs142616668 Chr.19: 45407183-45407183 on Build GRCh38

View Full Product Details