The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004961.3 | NP_004952.2 | 8 - 9 | 1191 | 76 |
XM_011531140.2 | XP_011529442.1 | 4 - 5 | 1648 | 76 | |
XM_017029386.1 | XP_016884875.1 | 8 - 9 | 2602 | 76 | |
XM_017029387.1 | XP_016884876.1 | 8 - 9 | 3243 | 76 | |
XM_017029388.1 | XP_016884877.1 | 4 - 5 | 575 | 76 | |
GenBank mRNA | AK303072.1 | - | 2 - 3 | 370 | 76 |
U66661.1 | - | 8 - 9 | 1177 | 76 | |
U92285.1 | - | 2 - 3 | 4216 | 76 | |
Y07637.1 | - | 8 - 9 | 1184 | 76 | |
Y09765.1 | - | 8 - 9 | 1191 | 76 |