Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105177165-105177394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
UniGene
Hs.506603
Gene Chromosome Location
Chr.12: 105173297-105236230 on Build GRCh38
Transcripts
NM_001251904.1 NM_001251905.1 NM_018171.3 XM_006719472.1 XM_011538530.2 XM_011538531.2 XM_011538532.2 XM_017019551.1 XM_017019552.1 XM_017019553.1 XM_017019554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1127584 c.1645C>T p.P549S Chr.12: 105177252-105177252 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373217394 Chr.12: 105177358-105177358 on Build GRCh38
rs756838524 Chr.12: 105177248-105177249 on Build GRCh38
rs778305467 Chr.12: 105177256-105177257 on Build GRCh38
rs767756992 Chr.12: 105177285-105177285 on Build GRCh38
rs368676151 Chr.12: 105177282-105177282 on Build GRCh38
rs935699948 Chr.12: 105177342-105177342 on Build GRCh38
rs749806205 Chr.12: 105177295-105177295 on Build GRCh38
rs757745361 Chr.12: 105177296-105177296 on Build GRCh38
rs751654637 Chr.12: 105177281-105177281 on Build GRCh38
rs773848624 Chr.12: 105177216-105177216 on Build GRCh38
rs761785692 Chr.12: 105177261-105177261 on Build GRCh38
rs375472756 Chr.12: 105177310-105177310 on Build GRCh38
rs745365981 Chr.12: 105177307-105177307 on Build GRCh38
rs757746014 Chr.12: 105177370-105177370 on Build GRCh38
rs758414805 Chr.12: 105177284-105177285 on Build GRCh38
rs374248068 Chr.12: 105177234-105177234 on Build GRCh38
rs996556192 Chr.12: 105177338-105177338 on Build GRCh38
rs73179990 Chr.12: 105177203-105177203 on Build GRCh38
rs763929438 Chr.12: 105177229-105177229 on Build GRCh38
rs745577754 Chr.12: 105177217-105177217 on Build GRCh38
rs369800359 Chr.12: 105177353-105177353 on Build GRCh38
rs753398882 Chr.12: 105177235-105177240 on Build GRCh38
rs770443632 Chr.12: 105177213-105177213 on Build GRCh38
rs778104566 Chr.12: 105177291-105177291 on Build GRCh38
rs764085045 Chr.12: 105177212-105177214 on Build GRCh38
rs745341740 Chr.12: 105177271-105177274 on Build GRCh38
rs189720343 Chr.12: 105177212-105177212 on Build GRCh38
rs775184236 Chr.12: 105177226-105177226 on Build GRCh38
rs185158969 Chr.12: 105177305-105177305 on Build GRCh38
rs1027729474 Chr.12: 105177373-105177373 on Build GRCh38
rs143283931 Chr.12: 105177247-105177247 on Build GRCh38
rs771729906 Chr.12: 105177225-105177225 on Build GRCh38
rs756501093 Chr.12: 105177289-105177289 on Build GRCh38
rs180760222 Chr.12: 105177270-105177270 on Build GRCh38
rs760574921 Chr.12: 105177227-105177227 on Build GRCh38
rs753899114 Chr.12: 105177253-105177256 on Build GRCh38
rs749078433 Chr.12: 105177361-105177361 on Build GRCh38
rs796315890 Chr.12: 105177365-105177366 on Build GRCh38
rs371633632 Chr.12: 105177287-105177287 on Build GRCh38
rs556287281 Chr.12: 105177326-105177326 on Build GRCh38
rs539224250 Chr.12: 105177301-105177301 on Build GRCh38
rs892083788 Chr.12: 105177372-105177372 on Build GRCh38
rs775196127 Chr.12: 105177359-105177359 on Build GRCh38
rs1029337347 Chr.12: 105177357-105177357 on Build GRCh38
rs766427688 Chr.12: 105177263-105177263 on Build GRCh38

View Full Product Details