The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010927.2 | NP_001010927.1 | 2 - 3 | 303 | 81 |
NM_012454.3 | NP_036586.2 | 15 - 16 | 3551 | 81 | |
GenBank mRNA | AF120323.1 | - | 11 - 12 | 1521 | 81 |
AF120324.1 | - | 2 - 3 | 279 | 81 | |
AK123859.1 | - | 16 - 17 | 3646 | 81 | |
AK125459.1 | - | 12 - 13 | 2010 | 81 | |
AK299723.1 | - | 2 - 3 | 300 | 81 | |
AK302860.1 | - | 11 - 12 | 1462 | 81 | |
AL122086.1 | - | 10 - 11 | 1380 | 81 | |
BC066979.1 | - | 2 - 3 | 300 | 81 | |
BC137275.1 | - | 14 - 15 | 3424 | 81 | |
BP199501.1 | - | 2 - 3 | 306 | 81 |