Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
Chromosome Location
Chr.13: 50909678 - 50970462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142279.2 NP_001135751.1 6 - 7 916 85
NM_024570.3 NP_078846.2 6 - 7 916 85
XM_006719867.3 XP_006719930.1 6 - 7 605 85
XM_011535229.1 XP_011533531.1 6 - 7 838 85
XM_011535230.2 XP_011533532.1 6 - 7 838 85
XM_011535231.2 XP_011533533.1 6 - 7 838 85
XM_011535233.2 XP_011533535.1 7 - 8 791 85
XM_017020747.1 XP_016876236.1 6 - 7 838 85
GenBank mRNA AK021774.1 - 6 - 7 803 85
AK124228.1 - 1 - 2 2582 85
AK223340.1 - 6 - 7 803 85
AY764036.1 - 6 - 7 824 85
BC001397.2 - 6 - 7 626 85
BC005088.1 - 6 - 7 626 85
BC007332.2 - 6 - 7 626 85
BC010174.2 - 6 - 7 626 85
BC036744.1 - 6 - 7 787 85
BX328027.1 - 3 - 4 243 85
BX330672.1 - 4 - 5 85

View Full Product Details