Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBA5
Entrez Gene ID
79876
Gene Name
ubiquitin like modifier activating enzyme 5
Gene Aliases
THIFP1, UBE1DC1
Chromosome Location
Chr.3: 132654446 - 132679786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024818.4 NP_079094.1 1 - 2 1202 69
GenBank mRNA AB154406.1 - 1 - 2 161 69
AK026904.1 - 1 - 2 395 69
AK056238.1 - 1 - 2 1203 69
AK129616.1 - 2 - 3 261 69
AL136757.1 - 1 - 2 215 69
AY253672.1 - 1 - 2 354 69
BC009737.1 - 1 - 2 331 69
DA785394.1 - 1 - 2 315 69
DB082596.1 - 1 - 2 404 69

View Full Product Details