Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161048973-161049230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3477115 c.2030G>A p.G677E Chr.1: 161048991-161048991 on Build GRCh38
898434 c.1891G>A p.G631R Chr.1: 161049130-161049130 on Build GRCh38
4981749 c.1855C>T p.L619F Chr.1: 161049166-161049166 on Build GRCh38
6122185 c.1897C>T p.L633L Chr.1: 161049124-161049124 on Build GRCh38
4533994 c.2038G>C p.E680Q Chr.1: 161048983-161048983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143582871 Chr.1: 161049090-161049090 on Build GRCh38
rs769634617 Chr.1: 161049199-161049199 on Build GRCh38
rs754561075 Chr.1: 161049059-161049059 on Build GRCh38
rs750300952 Chr.1: 161049034-161049034 on Build GRCh38
rs761631153 Chr.1: 161049021-161049021 on Build GRCh38
rs748364664 Chr.1: 161049192-161049192 on Build GRCh38
rs765180404 Chr.1: 161049028-161049028 on Build GRCh38
rs762914478 Chr.1: 161049205-161049205 on Build GRCh38
rs772339887 Chr.1: 161049078-161049078 on Build GRCh38
rs370782515 Chr.1: 161049082-161049082 on Build GRCh38
rs757766658 Chr.1: 161049145-161049145 on Build GRCh38
rs765919571 Chr.1: 161049109-161049109 on Build GRCh38
rs760795163 Chr.1: 161049011-161049011 on Build GRCh38
rs574520685 Chr.1: 161049017-161049017 on Build GRCh38
rs768722315 Chr.1: 161049187-161049187 on Build GRCh38
rs767093082 Chr.1: 161049130-161049130 on Build GRCh38
rs530186365 Chr.1: 161049045-161049045 on Build GRCh38
rs772503565 Chr.1: 161049202-161049202 on Build GRCh38
rs778236804 Chr.1: 161049061-161049061 on Build GRCh38
rs77801634 Chr.1: 161049072-161049072 on Build GRCh38
rs1030212838 Chr.1: 161049046-161049046 on Build GRCh38
rs775411304 Chr.1: 161049079-161049079 on Build GRCh38
rs750999799 Chr.1: 161049118-161049118 on Build GRCh38
rs552754298 Chr.1: 161049136-161049136 on Build GRCh38
rs747297606 Chr.1: 161049080-161049080 on Build GRCh38
rs759043726 Chr.1: 161049122-161049122 on Build GRCh38
rs201625032 Chr.1: 161049076-161049076 on Build GRCh38
rs749644105 Chr.1: 161049067-161049067 on Build GRCh38
rs531763065 Chr.1: 161049200-161049200 on Build GRCh38
rs375445197 Chr.1: 161049159-161049159 on Build GRCh38
rs1047866533 Chr.1: 161049022-161049022 on Build GRCh38
rs751279738 Chr.1: 161049053-161049053 on Build GRCh38
rs36089872 Chr.1: 161049106-161049106 on Build GRCh38
rs201140225 Chr.1: 161049020-161049020 on Build GRCh38
rs372513981 Chr.1: 161049107-161049107 on Build GRCh38
rs779570770 Chr.1: 161049075-161049075 on Build GRCh38
rs267598132 Chr.1: 161049004-161049004 on Build GRCh38
rs779412357 Chr.1: 161049151-161049151 on Build GRCh38
rs146914272 Chr.1: 161049165-161049165 on Build GRCh38
rs1006405683 Chr.1: 161049035-161049035 on Build GRCh38
rs781273702 Chr.1: 161049189-161049189 on Build GRCh38
rs757640016 Chr.1: 161049068-161049068 on Build GRCh38
rs775925131 Chr.1: 161049001-161049001 on Build GRCh38
rs747083587 Chr.1: 161049179-161049179 on Build GRCh38
rs765868638 Chr.1: 161049052-161049052 on Build GRCh38
rs368859663 Chr.1: 161049176-161049176 on Build GRCh38

View Full Product Details