The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000056.4 | NP_000047.1 | 7 - 8 | 926 | 88 |
NM_001318975.1 | NP_001305904.1 | 7 - 8 | 720 | 88 | |
NM_183050.3 | NP_898871.1 | 7 - 8 | 926 | 88 | |
NR_134945.1 | 8 - 9 | 1020 | 88 | ||
XM_005248756.4 | XP_005248813.1 | 7 - 8 | 875 | 88 | |
XM_011536023.2 | XP_011534325.1 | 7 - 8 | 875 | 88 | |
XM_011536024.2 | XP_011534326.1 | 7 - 8 | 875 | 88 | |
XM_011536025.2 | XP_011534327.1 | 7 - 8 | 875 | 88 | |
XM_011536026.2 | XP_011534328.1 | 7 - 8 | 862 | 88 | |
GenBank mRNA | AK289977.1 | - | 7 - 8 | 855 | 88 |
AK304353.1 | - | 7 - 8 | 691 | 88 | |
AK310838.1 | - | 5 - 6 | 684 | 88 | |
AK316545.1 | - | 8 - 9 | 839 | 88 | |
BC034481.1 | - | 8 - 9 | 964 | 88 | |
BC040139.2 | - | 7 - 8 | 847 | 88 | |
BM553932.1 | - | 3 - 4 | 283 | 88 | |
BT020063.1 | - | 7 - 8 | 842 | 88 | |
M55575.1 | - | 7 - 8 | 846 | 88 | |
U50708.1 | - | 7 - 8 | 889 | 88 | |
X52446.1 | - | 7 - 8 | 785 | 88 |