The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000164.3 | NP_000155.1 | 7 - 8 | 736 | 114 |
NM_001308418.1 | NP_001295347.1 | 6 - 7 | 628 | 114 | |
XM_011526709.2 | XP_011525011.1 | 7 - 8 | 793 | 114 | |
XM_011526710.2 | XP_011525012.1 | 8 - 9 | 874 | 114 | |
XM_011526711.2 | XP_011525013.1 | 6 - 7 | 685 | 114 | |
XM_011526714.2 | XP_011525016.1 | 4 - 5 | 469 | 114 | |
XM_011526715.2 | XP_011525017.1 | 4 - 5 | 479 | 114 | |
XM_011526716.2 | XP_011525018.1 | 7 - 8 | 793 | 114 | |
GenBank mRNA | BC093723.1 | - | 7 - 8 | 723 | 114 |
BC095494.1 | - | 6 - 7 | 635 | 114 | |
BC101673.1 | - | 7 - 8 | 723 | 114 | |
BC143300.1 | - | 6 - 7 | 616 | 114 | |
BC143302.1 | - | 7 - 8 | 723 | 114 | |
S79852.1 | - | 7 - 8 | 731 | 114 | |
U39231.1 | - | 7 - 8 | 736 | 114 |