The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001560.2 | NP_001551.1 | 10 - 11 | 1231 | 106 |
XM_011531336.1 | XP_011529638.1 | 10 - 11 | 1509 | 106 | |
XM_011531337.1 | XP_011529639.1 | 3 - 4 | 1538 | 106 | |
XM_017029507.1 | XP_016884996.1 | 9 - 10 | 1248 | 106 | |
GenBank mRNA | AB209849.1 | - | 10 - 11 | 1186 | 106 |
AK123515.1 | - | 3 - 4 | 1194 | 106 | |
AK296958.1 | - | 5 - 6 | 460 | 106 | |
AK313467.1 | - | 10 - 11 | 1220 | 106 | |
BC009960.2 | - | 10 - 11 | 1218 | 106 | |
BC015768.1 | - | 10 - 11 | 1235 | 106 | |
U81379.3 | - | 10 - 11 | 1228 | 106 | |
Y09328.1 | - | 10 - 11 | 1221 | 106 | |
Y10659.1 | - | 10 - 11 | 1231 | 106 |