The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002224.3 | NP_002215.2 | 12 - 13 | 1478 | 98 |
XM_011514577.2 | XP_011512879.1 | 11 - 12 | 1118 | 98 | |
XM_017010832.1 | XP_016866321.1 | 13 - 14 | 1307 | 98 | |
GenBank mRNA | AB209739.1 | - | 3 - 4 | 265 | 98 |
BC146646.1 | - | 12 - 13 | 1437 | 98 | |
D26351.1 | - | 12 - 13 | 1489 | 98 | |
U01062.1 | - | 12 - 13 | 1290 | 98 |