The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003181.3 | NP_003172.1 | 7 - 8 | 1424 | 67 |
XM_011536080.2 | XP_011534382.1 | 6 - 7 | 1194 | 67 | |
GenBank mRNA | AJ001699.1 | - | 6 - 7 | 1064 | 67 |
BC039877.1 | - | 6 - 7 | 1194 | 67 |