Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAV2
Entrez Gene ID
7410
Gene Name
vav guanine nucleotide exchange factor 2
Gene Aliases
VAV-2
Chromosome Location
Chr.9: 133761894 - 133992374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134398.1 NP_001127870.1 10 - 11 976 79
NM_003371.3 NP_003362.2 9 - 10 961 79
XM_005272213.1 XP_005272270.1 9 - 10 1011 79
XM_017015108.1 XP_016870597.1 10 - 11 1026 79
XM_017015109.1 XP_016870598.1 9 - 10 1011 79
XM_017015110.1 XP_016870599.1 10 - 11 1026 79
XM_017015111.1 XP_016870600.1 10 - 11 1026 79
XM_017015112.1 XP_016870601.1 10 - 11 1026 79
XM_017015113.1 XP_016870602.1 10 - 11 1026 79
GenBank mRNA BC132965.1 - 9 - 10 961 79
BC132967.1 - 9 - 10 961 79
BQ050947.1 - 5 - 6 415 79
BX640754.1 - 9 - 10 801 79
S76992.1 - 10 - 11 935 79

View Full Product Details