Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 14 - 15 2692 81
NM_001322988.1 NP_001309917.1 14 - 15 2692 81
NM_001322989.1 NP_001309918.1 14 - 15 2692 81
NM_001322990.1 NP_001309919.1 14 - 15 2653 81
NM_001322991.1 NP_001309920.1 14 - 15 2653 81
NM_001322992.1 NP_001309921.1 14 - 15 2692 81
NM_003947.5 NP_003938.1 14 - 15 2692 81
XM_006713810.3 XP_006713873.1 14 - 15 2937 81
XM_006713811.3 XP_006713874.1 14 - 15 2937 81
XM_006713812.3 XP_006713875.1 14 - 15 2898 81
XM_006713813.3 XP_006713876.1 14 - 15 2898 81
XM_006713814.3 XP_006713877.1 14 - 15 2937 81
XM_006713815.3 XP_006713878.1 14 - 15 2940 81
XM_011513279.2 XP_011511581.1 14 - 15 2937 81
XM_011513280.2 XP_011511582.1 14 - 15 2938 81
XM_011513281.2 XP_011511583.1 14 - 15 25586 81
XM_011513283.2 XP_011511585.1 14 - 15 2934 81
XM_011513285.2 XP_011511587.1 14 - 15 2933 81
XM_017007429.1 XP_016862918.1 14 - 15 2937 81
XM_017007430.1 XP_016862919.1 14 - 15 2937 81
XM_017007431.1 XP_016862920.1 14 - 15 2933 81
XM_017007432.1 XP_016862921.1 14 - 15 2933 81
XM_017007433.1 XP_016862922.1 14 - 15 2940 81
XM_017007434.1 XP_016862923.1 14 - 15 2901 81
XM_017007435.1 XP_016862924.1 14 - 15 2901 81
GenBank mRNA AK131379.1 - 4 - 5 753 81
AK314126.1 - 14 - 15 2692 81
U94190.1 - 14 - 15 2635 81

View Full Product Details