The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 3 - 4 | 611 | 61 |
XM_006718056.3 | XP_006718119.1 | 3 - 4 | 574 | 61 | |
XM_006718057.3 | XP_006718120.1 | 3 - 4 | 581 | 61 | |
XM_011540341.2 | XP_011538643.1 | 3 - 4 | 735 | 61 | |
XM_011540342.1 | XP_011538644.1 | 3 - 4 | 313 | 61 | |
XM_011540344.2 | XP_011538646.1 | 2 - 3 | 220 | 61 | |
XM_011540345.1 | XP_011538647.1 | 2 - 3 | 345 | 61 | |
XM_011540346.2 | XP_011538648.1 | 3 - 4 | 580 | 61 | |
XM_017016913.1 | XP_016872402.1 | 3 - 4 | 274 | 61 | |
XM_017016914.1 | XP_016872403.1 | 3 - 4 | 604 | 61 | |
GenBank mRNA | AB011155.2 | - | 3 - 4 | 641 | 61 |
AB091676.1 | - | 1 - 2 | 76 | 61 | |
AK122931.1 | - | 3 - 4 | 316 | 61 | |
BC146794.1 | - | 3 - 4 | 641 | 61 |