The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 8 - 9 | 1691 | 65 |
XM_005270276.4 | XP_005270333.1 | 8 - 9 | 1645 | 65 | |
XM_006718056.3 | XP_006718119.1 | 8 - 9 | 1654 | 65 | |
XM_006718057.3 | XP_006718120.1 | 8 - 9 | 1661 | 65 | |
XM_011540341.2 | XP_011538643.1 | 8 - 9 | 1815 | 65 | |
XM_011540342.1 | XP_011538644.1 | 8 - 9 | 1393 | 65 | |
XM_011540344.2 | XP_011538646.1 | 7 - 8 | 1300 | 65 | |
XM_011540345.1 | XP_011538647.1 | 7 - 8 | 1425 | 65 | |
XM_011540346.2 | XP_011538648.1 | 8 - 9 | 1660 | 65 | |
XM_011540347.2 | XP_011538649.1 | 4 - 5 | 871 | 65 | |
XM_017016913.1 | XP_016872402.1 | 8 - 9 | 1354 | 65 | |
XM_017016914.1 | XP_016872403.1 | 8 - 9 | 1684 | 65 | |
XM_017016915.1 | XP_016872404.1 | 4 - 5 | 1555 | 65 | |
GenBank mRNA | AB011155.2 | - | 8 - 9 | 1721 | 65 |
AB091676.1 | - | 6 - 7 | 1156 | 65 | |
AK055172.1 | - | 4 - 5 | 609 | 65 | |
AK122931.1 | - | 9 - 10 | 1623 | 65 | |
BC146794.1 | - | 8 - 9 | 1721 | 65 | |
BX649131.1 | - | 4 - 5 | 818 | 65 |