The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004899.4 | NP_004890.2 | 12 - 13 | 1513 | 70 |
GenBank mRNA | AK000097.1 | - | 12 - 13 | 1399 | 70 |
AK098345.1 | - | 1 - 2 | 1431 | 70 | |
CR994399.1 | - | 2 - 3 | 255 | 70 |