Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
Chromosome Location
Chr.21: 44107262 - 44131181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005049.2 NP_005040.2 12 - 13 1570 65
XM_011529667.2 XP_011527969.1 12 - 13 1635 65
GenBank mRNA AB209438.1 - 3 - 4 657 65
AK314184.1 - 12 - 13 1574 65
BC013309.2 - 12 - 13 1480 65
BC014988.1 - 12 - 13 1480 65
BM457858.1 - 3 - 4 403 65
U53346.1 - 12 - 13 1504 65
U56085.1 - 12 - 13 1515 65
X95263.1 - 12 - 13 1511 65

Gene Symbol
LOC102724159
Entrez Gene ID
102724159
Gene Name
periodic tryptophan protein 2 homolog
Gene Aliases
-
Chromosome Location
Chr.21: 5130871 - 5154767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006723910.1 XP_006723973.1 12 - 13 1607 65

View Full Product Details