Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
Chromosome Location
Chr.21: 44107262 - 44131181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005049.2 NP_005040.2 18 - 19 2447 114
GenBank mRNA AK314184.1 - 18 - 19 2451 114
BC013309.2 - 18 - 19 2357 114
BC014988.1 - 18 - 19 2357 114
BX387451.2 - 6 - 7 757 114
U53346.1 - 18 - 19 2381 114
U56085.1 - 18 - 19 2392 114
X95263.1 - 18 - 19 2388 114

Gene Symbol
LOC102724159
Entrez Gene ID
102724159
Gene Name
periodic tryptophan protein 2 homolog
Gene Aliases
-
Chromosome Location
Chr.21: 5130871 - 5154767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006723910.1 XP_006723973.1 18 - 19 2484 114

View Full Product Details