Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
Chromosome Location
Chr.21: 44107262 - 44131181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005049.2 NP_005040.2 9 - 10 1143 108
XM_011529667.2 XP_011527969.1 9 - 10 1208 108
GenBank mRNA AK314184.1 - 9 - 10 1147 108
BC013309.2 - 9 - 10 1053 108
BC014988.1 - 9 - 10 1053 108
BX452317.1 - 4 - 5 499 108
U53346.1 - 9 - 10 1077 108
U56085.1 - 9 - 10 1088 108
X95263.1 - 9 - 10 1084 108

Gene Symbol
LOC102724159
Entrez Gene ID
102724159
Gene Name
periodic tryptophan protein 2 homolog
Gene Aliases
-
Chromosome Location
Chr.21: 5130871 - 5154767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006723910.1 XP_006723973.1 9 - 10 1180 108

View Full Product Details