The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031685.2 | NP_001026855.2 | 8 - 9 | 1283 | 137 |
NM_005426.2 | NP_005417.1 | 9 - 10 | 1415 | 137 | |
XM_011544268.1 | XP_011542570.1 | 7 - 8 | 1144 | 137 | |
XM_011544269.2 | XP_011542571.1 | 5 - 6 | 802 | 137 | |
XM_017002220.1 | XP_016857709.1 | 9 - 10 | 1424 | 137 | |
GenBank mRNA | AJ318888.2 | - | 8 - 9 | 1228 | 137 |
AK294432.1 | - | 8 - 9 | 1135 | 137 | |
AK295378.1 | - | 1 - 2 | 61 | 137 | |
BC040247.1 | - | 6 - 7 | 759 | 137 | |
BC046150.1 | - | 8 - 9 | 1226 | 137 | |
BC058918.1 | - | 8 - 9 | 1241 | 137 | |
U58334.1 | - | 9 - 10 | 1360 | 137 |