The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007235.4 | NP_009166.2 | 2 - 3 | 564 | 104 |
XM_017018748.1 | XP_016874237.1 | 2 - 3 | 424 | 104 | |
GenBank mRNA | AF039022.1 | - | 1 - 2 | 58 | 104 |
AK292884.1 | - | 2 - 3 | 377 | 104 | |
AK308108.1 | - | 2 - 3 | 385 | 104 | |
BC020569.1 | - | 1 - 2 | 143 | 104 | |
BQ215290.1 | - | 2 - 3 | 484 | 104 | |
CR979468.1 | - | 2 - 3 | 564 | 104 | |
Y16414.1 | - | 1 - 2 | 116 | 104 |