The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007243.2 | NP_009174.1 | 3 - 4 | 683 | 89 |
NR_073065.1 | 2 - 3 | 508 | 111 | ||
XM_017010224.1 | XP_016865713.1 | 2 - 3 | 685 | 89 | |
XM_017010225.1 | XP_016865714.1 | 3 - 4 | 599 | 89 | |
GenBank mRNA | AF143676.1 | - | 2 - 3 | 329 | 89 |
AK075509.1 | - | 3 - 4 | 685 | 89 | |
AK315190.1 | - | 2 - 3 | 341 | 89 | |
AY358411.1 | - | 2 - 3 | 350 | 89 | |
BC021260.1 | - | 2 - 3 | 321 | 89 | |
BC039865.1 | - | 2 - 3 | 349 | 89 | |
BE548635.1 | - | 2 - 3 | 328 | 89 | |
BM847576.1 | - | 2 - 3 | 382 | 111 |