The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195733.1 | NP_001182662.1 | 1 - 2 | 179 | 54 |
NM_001300849.1 | NP_001287778.1 | 1 - 2 | 192 | 54 | |
NM_012398.2 | NP_036530.1 | 1 - 2 | 179 | 54 | |
XM_011527845.1 | XP_011526147.1 | 1 - 2 | 186 | 54 | |
XM_011527846.1 | XP_011526148.1 | 1 - 2 | 186 | 54 | |
XM_011527848.1 | XP_011526150.1 | 1 - 2 | 186 | 54 | |
XM_011527849.1 | XP_011526151.1 | 1 - 2 | 186 | 54 | |
XM_017026540.1 | XP_016882029.1 | 1 - 2 | 186 | 54 | |
GenBank mRNA | AB011161.1 | - | 1 - 2 | 147 | 54 |
AK301309.1 | - | 1 - 2 | 162 | 54 | |
AK315912.1 | - | 1 - 2 | 165 | 54 | |
FJ965536.1 | - | 1 - 2 | 90 | 54 | |
FJ965537.1 | - | 1 - 2 | 90 | 54 | |
HY000479.1 | - | 1 - 2 | 192 | 54 |