The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032319.1 | NP_115695.1 | 3 - 4 | 346 | 64 |
GenBank mRNA | AK312077.1 | - | 3 - 4 | 354 | 64 |
AY358605.1 | - | 3 - 4 | 337 | 64 | |
BC005069.1 | - | 3 - 4 | 346 | 64 |