Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
Chromosome Location
Chr.17: 28394642 - 28406630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080669.5 NP_542400.2 3 - 4 1270 72
XM_017024110.1 XP_016879599.1 4 - 5 1229 72
GenBank mRNA AK054669.1 - 3 - 4 1268 72
AL832613.2 - 2 - 3 578 72

View Full Product Details