Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT1C2P1
Entrez Gene ID
151234
Gene Name
sulfotransferase family 1C member 2 pseudogene 1
Gene Aliases
SULT1C1P
Chromosome Location
Chr.2: 108322238 - 108353799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037191.1 3 - 4 452 88
GenBank mRNA AK056906.1 - 3 - 4 452 88

View Full Product Details