Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202035998-202036269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
UniGene
Hs.173859
Gene Chromosome Location
Chr.2: 202034587-202038437 on Build GRCh38
Transcripts
NM_003507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668179 c.1452C>T p.I484I Chr.2: 202036099-202036099 on Build GRCh38
6668180 c.1537G>A p.V513M Chr.2: 202036184-202036184 on Build GRCh38
6668178 c.1409G>A p.R470H Chr.2: 202036056-202036056 on Build GRCh38
7143432 c.1605G>A p.L535L Chr.2: 202036252-202036252 on Build GRCh38
5037268 c.1510C>A p.L504I Chr.2: 202036157-202036157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000638888 Chr.2: 202036030-202036030 on Build GRCh38
rs762632983 Chr.2: 202036128-202036128 on Build GRCh38
rs182592123 Chr.2: 202036195-202036195 on Build GRCh38
rs946938708 Chr.2: 202036205-202036205 on Build GRCh38
rs891140187 Chr.2: 202036209-202036209 on Build GRCh38
rs1038885248 Chr.2: 202036090-202036090 on Build GRCh38
rs775589753 Chr.2: 202036126-202036126 on Build GRCh38
rs201452450 Chr.2: 202036118-202036118 on Build GRCh38
rs763782149 Chr.2: 202036214-202036214 on Build GRCh38
rs763985883 Chr.2: 202036129-202036129 on Build GRCh38
rs200370632 Chr.2: 202036186-202036186 on Build GRCh38
rs746743333 Chr.2: 202036087-202036087 on Build GRCh38
rs146693247 Chr.2: 202036020-202036020 on Build GRCh38
rs761598219 Chr.2: 202036231-202036231 on Build GRCh38
rs774136031 Chr.2: 202036223-202036223 on Build GRCh38
rs749570044 Chr.2: 202036031-202036031 on Build GRCh38
rs143528185 Chr.2: 202036183-202036183 on Build GRCh38
rs772709570 Chr.2: 202036070-202036070 on Build GRCh38
rs767737659 Chr.2: 202036162-202036162 on Build GRCh38
rs769473710 Chr.2: 202036203-202036203 on Build GRCh38
rs541873049 Chr.2: 202036182-202036182 on Build GRCh38
rs751432827 Chr.2: 202036178-202036179 on Build GRCh38
rs770553279 Chr.2: 202036097-202036097 on Build GRCh38
rs997348548 Chr.2: 202036141-202036141 on Build GRCh38
rs1029625624 Chr.2: 202036172-202036172 on Build GRCh38
rs780572956 Chr.2: 202036029-202036029 on Build GRCh38
rs35600847 Chr.2: 202036107-202036107 on Build GRCh38
rs775255613 Chr.2: 202036204-202036204 on Build GRCh38
rs368422200 Chr.2: 202036159-202036159 on Build GRCh38
rs372525407 Chr.2: 202036174-202036174 on Build GRCh38
rs745320558 Chr.2: 202036198-202036198 on Build GRCh38
rs140867221 Chr.2: 202036238-202036238 on Build GRCh38
rs772230105 Chr.2: 202036052-202036052 on Build GRCh38
rs770643171 Chr.2: 202036192-202036192 on Build GRCh38
rs767678664 Chr.2: 202036237-202036237 on Build GRCh38
rs776225705 Chr.2: 202036099-202036099 on Build GRCh38
rs750464553 Chr.2: 202036168-202036168 on Build GRCh38
rs765225022 Chr.2: 202036117-202036117 on Build GRCh38
rs562818320 Chr.2: 202036150-202036150 on Build GRCh38
rs750694306 Chr.2: 202036246-202036246 on Build GRCh38
rs759120037 Chr.2: 202036103-202036103 on Build GRCh38
rs763013166 Chr.2: 202036207-202036207 on Build GRCh38
rs1004437982 Chr.2: 202036222-202036222 on Build GRCh38
rs201191053 Chr.2: 202036064-202036064 on Build GRCh38

View Full Product Details