Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44825374-44825625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP8
Entrez Gene ID
23779
Gene Name
Rho GTPase activating protein 8
Gene Aliases
BPGAP1, PP610
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44752558-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
271486 c.653C>A p.P218Q Chr.22: 44825542-44825542 on Build GRCh38
3973004 c.691G>A p.V231I Chr.22: 44825580-44825580 on Build GRCh38
7235852 c.614G>A p.R205Q Chr.22: 44825518-44825518 on Build GRCh38
5513731 c.663G>A p.R221R Chr.22: 44825552-44825552 on Build GRCh38
5818754 c.648G>T p.R216R Chr.22: 44825552-44825552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761677722 Chr.22: 44825591-44825591 on Build GRCh38
rs973882621 Chr.22: 44825393-44825393 on Build GRCh38
rs1044642170 Chr.22: 44825483-44825483 on Build GRCh38
rs773455283 Chr.22: 44825522-44825522 on Build GRCh38
rs374830968 Chr.22: 44825452-44825452 on Build GRCh38
rs375415792 Chr.22: 44825533-44825533 on Build GRCh38
rs745573105 Chr.22: 44825445-44825445 on Build GRCh38
rs377295717 Chr.22: 44825549-44825549 on Build GRCh38
rs768402468 Chr.22: 44825589-44825589 on Build GRCh38
rs758070305 Chr.22: 44825511-44825511 on Build GRCh38
rs769217037 Chr.22: 44825462-44825462 on Build GRCh38
rs761975823 Chr.22: 44825470-44825470 on Build GRCh38
rs1036661085 Chr.22: 44825425-44825425 on Build GRCh38
rs114896130 Chr.22: 44825428-44825428 on Build GRCh38
rs752551284 Chr.22: 44825482-44825482 on Build GRCh38
rs753817080 Chr.22: 44825489-44825489 on Build GRCh38
rs184112239 Chr.22: 44825429-44825429 on Build GRCh38
rs576293386 Chr.22: 44825423-44825423 on Build GRCh38
rs1036392504 Chr.22: 44825526-44825526 on Build GRCh38
rs35480376 Chr.22: 44825604-44825605 on Build GRCh38
rs909574683 Chr.22: 44825415-44825415 on Build GRCh38
rs1029048490 Chr.22: 44825403-44825403 on Build GRCh38
rs949927755 Chr.22: 44825504-44825504 on Build GRCh38
rs370649393 Chr.22: 44825547-44825547 on Build GRCh38
rs776418093 Chr.22: 44825590-44825590 on Build GRCh38
rs34702238 Chr.22: 44825573-44825574 on Build GRCh38
rs74393756 Chr.22: 44825524-44825524 on Build GRCh38
rs371358036 Chr.22: 44825432-44825432 on Build GRCh38
rs146040774 Chr.22: 44825521-44825521 on Build GRCh38
rs778419615 Chr.22: 44825502-44825502 on Build GRCh38
rs376772948 Chr.22: 44825545-44825545 on Build GRCh38
rs747404579 Chr.22: 44825459-44825459 on Build GRCh38
rs763930138 Chr.22: 44825537-44825537 on Build GRCh38
rs369911894 Chr.22: 44825535-44825535 on Build GRCh38
rs780222524 Chr.22: 44825513-44825513 on Build GRCh38
rs368897846 Chr.22: 44825510-44825510 on Build GRCh38
rs765855179 Chr.22: 44825606-44825606 on Build GRCh38
rs117749664 Chr.22: 44825518-44825518 on Build GRCh38
rs369660678 Chr.22: 44825517-44825517 on Build GRCh38
rs771172363 Chr.22: 44825525-44825525 on Build GRCh38
rs765143772 Chr.22: 44825597-44825597 on Build GRCh38
rs759702968 Chr.22: 44825583-44825583 on Build GRCh38
rs3209476 Chr.22: 44825543-44825543 on Build GRCh38
rs533325275 Chr.22: 44825514-44825514 on Build GRCh38
rs140257857 Chr.22: 44825580-44825580 on Build GRCh38
rs953896160 Chr.22: 44825541-44825541 on Build GRCh38
rs750390216 Chr.22: 44825604-44825604 on Build GRCh38
rs770428449 Chr.22: 44825557-44825557 on Build GRCh38
rs781706922 Chr.22: 44825552-44825552 on Build GRCh38
rs763256370 Chr.22: 44825476-44825476 on Build GRCh38
rs777330904 Chr.22: 44825465-44825465 on Build GRCh38
rs549994660 Chr.22: 44825515-44825515 on Build GRCh38
rs756742080 Chr.22: 44825494-44825494 on Build GRCh38
rs200605462 Chr.22: 44825454-44825454 on Build GRCh38
rs865889338 Chr.22: 44825407-44825407 on Build GRCh38
rs151197082 Chr.22: 44825546-44825546 on Build GRCh38
rs540758148 Chr.22: 44825473-44825473 on Build GRCh38
rs773545359 Chr.22: 44825474-44825474 on Build GRCh38
rs770956991 Chr.22: 44825579-44825579 on Build GRCh38
rs753730992 Chr.22: 44825539-44825539 on Build GRCh38
rs137928246 Chr.22: 44825414-44825414 on Build GRCh38
rs748623441 Chr.22: 44825554-44825554 on Build GRCh38
rs755347776 Chr.22: 44825551-44825551 on Build GRCh38
rs778484303 Chr.22: 44825558-44825558 on Build GRCh38
rs546068174 Chr.22: 44825402-44825402 on Build GRCh38
rs189531344 Chr.22: 44825605-44825605 on Build GRCh38
rs565896716 Chr.22: 44825548-44825548 on Build GRCh38
rs144191237 Chr.22: 44825486-44825486 on Build GRCh38
rs767175171 Chr.22: 44825479-44825479 on Build GRCh38

View Full Product Details