Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14852129-14852402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
UniGene
Hs.534169
Gene Chromosome Location
Chr.10: 14838160-14871741 on Build GRCh38
Transcripts
NM_016299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7128795 c.574A>C p.N192H Chr.10: 14852371-14852371 on Build GRCh38
4804376 c.579T>C p.I193I Chr.10: 14852376-14852376 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs78383693 Chr.10: 14852180-14852180 on Build GRCh38
rs954503242 Chr.10: 14852330-14852330 on Build GRCh38
rs143776123 Chr.10: 14852337-14852337 on Build GRCh38
rs747213066 Chr.10: 14852195-14852195 on Build GRCh38
rs377647045 Chr.10: 14852340-14852340 on Build GRCh38
rs760525969 Chr.10: 14852347-14852347 on Build GRCh38
rs779955618 Chr.10: 14852356-14852356 on Build GRCh38
rs974391267 Chr.10: 14852241-14852241 on Build GRCh38
rs370840900 Chr.10: 14852349-14852349 on Build GRCh38
rs763834442 Chr.10: 14852333-14852339 on Build GRCh38
rs376227646 Chr.10: 14852332-14852332 on Build GRCh38
rs748234226 Chr.10: 14852338-14852338 on Build GRCh38
rs964227253 Chr.10: 14852235-14852235 on Build GRCh38
rs901950613 Chr.10: 14852190-14852190 on Build GRCh38
rs773835852 Chr.10: 14852345-14852349 on Build GRCh38
rs776384191 Chr.10: 14852352-14852352 on Build GRCh38
rs951566776 Chr.10: 14852162-14852162 on Build GRCh38
rs113350106 Chr.10: 14852266-14852266 on Build GRCh38
rs576506930 Chr.10: 14852343-14852343 on Build GRCh38

View Full Product Details